MilliporeSigma
All Photos(1)

S3639

Sigma-Aldrich

[D-Arg1, D-Pro2, D-Phe7, D-His9]-Substance P

≥95% (HPLC)

Synonym(s):
D-Arg-D-Pro-Lys-Pro-Gln-Gln-D-Phe-Phe-D-His-Leu-Met-NH2
Empirical Formula (Hill Notation):
C67H102N20O13S
CAS Number:
Molecular Weight:
1427.72
MDL number:

assay

≥95% (HPLC)

composition

Peptide content, ~70%

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)C(Cc1cnc[nH]1)NC(=O)C(Cc2ccccc2)NC(=O)C(Cc3ccccc3)NC(=O)C(CCC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C4CCCN4C(=O)C(CCCCN)NC(=O)C5CCCN5C(=O)C(N)CCCNC(N)=N)C(N)=O

Biochem/physiol Actions

Substance P antagonist

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service