MilliporeSigma
All Photos(1)

S4262

Sigma-Aldrich

Sera from goose

lyophilized powder

biological source

animal (goose)

form

lyophilized powder

storage temp.

2-8°C

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service