S6564

Sigma-Aldrich

SQI-PR sodium form

≥90% (TLC)

Empirical Formula (Hill Notation):
C42H59N4O7
Molecular Weight:
731.94
MDL number:

assay

≥90% (TLC)

storage temp.

−20°C

SMILES string

CCCOc1cc(N2CCOCCOCCN(CCOCC2)c3cc(OCCC)c(OCCC)c4ccc(C)nc34)c5nc(C)ccc5c1OCCC

Application

Sodium ionophore with a selectivity of 100:1 for sodium over potassium. Effective at concentrations of 1-5 μM.

Physical form

Orange-brown gum

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service