MilliporeSigma
All Photos(1)

S6636

Sigma-Aldrich

[Sar9]-Substance P

≥97% (HPLC)

Synonym(s):
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Sar-Leu-Met-NH2
Empirical Formula (Hill Notation):
C64H100N18O13S
CAS Number:
Molecular Weight:
1361.66
MDL number:
PubChem Substance ID:

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)CN(C)C(=O)C(Cc1ccccc1)NC(=O)C(Cc2ccccc2)NC(=O)C(CCC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C3CCCN3C(=O)C(CCCCN)NC(=O)C4CCCN4C(=O)C(N)CCCNC(N)=N)C(N)=O

Biochem/physiol Actions

NK-1 receptor agonist.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service