SAB1100364

Sigma-Aldrich

Anti-PFKL (361-375) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):
Anti-Phosphofructokinase, liver, Anti-PFK-B
Pricing and availability is not currently available.

biological source

rabbit

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~85 kDa

species reactivity

human

application(s)

western blot: 1:500-1:2,000

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PFKL(5211)

Immunogen

synthetic peptide corresponding to amino acids 361-375 of human PFKL

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

RIDADR

NONH for all modes of transport

WGK Germany

2

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.