MilliporeSigma
All Photos(1)

SAB1101280

Sigma-Aldrich

Anti-GORASP2 (43-55) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):
Anti-GOLPH6, Anti-GRASP55, Anti-Golgi reassembly stacking protein 2, 55kDa, Anti-p59
MDL number:

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~47 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GORASP2(26003)

Immunogen

synthetic peptide corresponding to amino acids 43-55 of human GORASP2

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service