All Photos(1)

SAB1101874

Sigma-Aldrich

Anti-KLC2 (421-435) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):
Anti-Kinesin light chain 2

biological source

rabbit

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~69 kDa

species reactivity

human

application(s)

western blot: 1:500-1:2,000

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... KLC2(64837)

Immunogen

synthetic peptide corresponding to amino acids 421-435 of human KLC2

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service