MilliporeSigma
All Photos(1)

SAB1101903

Sigma-Aldrich

Anti-A2BP1 (71-85) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):
Anti-Ataxin 2-binding protein 1, Anti-FOX1, Anti-HRNBP1

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~43 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... A2BP1(54715)

Immunogen

synthetic peptide corresponding to amino acids 71-85 of human A2BP1

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service