SAB2100160

Sigma-Aldrich

Anti-ASAHL antibody produced in rabbit

affinity isolated antibody

Synonym(s):
Anti-NAAA, Anti-PLT, Anti-ASAHL

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

37 kDa

species reactivity

human, bovine, rabbit, rat, guinea pig, horse, mouse

concentration

0.5 mg - 1 mg/mL

application(s)

western blot: suitable

conjugate

unconjugated

storage temp.

−20°C

Gene Information

human ... ASAHL(27163)

Immunogen

The immunogen for anti-ASAHL antibody: synthetic peptide derected towards the middle region of human ASAHL

Sequence

Synthetic peptide located within the following region: VSWLIRATLSESENFEAAVGKLAKTPLIADVYYIVGGTSPREGVVITRNR

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Rezan Abdul-Kadir et al.
Transfusion, 54(7), 1756-1768 (2014-03-13)
Postpartum hemorrhage (PPH) remains one of the leading causes of maternal morbidity and mortality worldwide, although the lack of a precise definition precludes accurate data of the absolute prevalence of PPH. An international expert panel in obstetrics, gynecology, hematology, transfusion...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service