SHC011

Sigma-Aldrich

MISSION® pLKO.1-puro-CMV-TagYFP Positive Control Plasmid DNA

Contains a gene encoding TagYFP

Synonym(s):
MISSION Control Vectors
MDL number:
Pricing and availability is not currently available.

product line

MISSION®

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells respectively. In addition, we recommend producing self-inactivating replication incompetent viral particles in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. The CMV-TagYFP Control Vector is provided as 10 μg of plasmid DNA in Tris-EDTA (TE) buffer at a concentration of 500 ng/μl.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.
The MISSION TagYFP Control Vector is an 8599 base pair lentivirus plasmid vector that contains a gene encoding TagYFP, under the control of the CMV promoter. The CMV-TagYFP Control Vector is useful as a positive control in experiments using the MISSION shRNA library clones.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense.
MISSION is a registered trademark of Sigma-Aldrich Co. LLC
TagYFP is a trademark of Evrogen Co.

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.