SHC013V

Sigma-Aldrich

MISSION® pLKO.1-puro-CMV-TagFP635 Positive Control Transduction Particles

Contains a gene encoding TagFP635

Synonym(s):
MISSION TurboGFP Control Transduction Particles
Pricing and availability is not currently available.

product line

MISSION®

application(s)

capture ELISA: 106 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. In addition, the Control Transduction Particles are pseudotyped with an envelope G glycoprotein from Vesicular Stomatitis Virus (VSV-G), allowing transduction of a wide variety of mammalian cells. 200 μl of 106 TU/ml (via p24 titering assay) lentiviral particles are provided as frozen stock.
When conducting experiments using MISSION® shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION Control Transduction Particles are a critical positive control to monitor transduction efficiency.
To see more application data, protocols, vector maps visit
sigma.com/shrna.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.
When conducting experiments using MISSION shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION TagFP635 Control Transduction Particles are a positive control to monitor transduction efficiency. The MISSION TagFP635 Control Transduction Particles are produced from the sequence-verified 8593 base pair lentivirus plasmid, pLKO.1-puro-CMV-TagFP635 (SHC013).

This construct aids in interpretation of experimental design and results by providing a far-red fluorescent protein marker for monitoring success in transduction of your cells of interest.

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC
TagFP635 is a trademark of Evrogen Co.

RIDADR

UN3373 - class 6.2 Biological substance, Category B

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.