T4658

Sigma-Aldrich

Tetrahexylammonium chloride

Linear Formula:
[CH3(CH2)5]4N(Cl)
CAS Number:
Molecular Weight:
390.13
Beilstein/REAXYS Number:
4605091
EC Number:
MDL number:

mp

111-113 °C (lit.)

SMILES string

[Cl-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.ClH/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;/h5-24H2,1-4H3;1H/q+1;/p-1

InChI key

ODTSDWCGLRVBHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service