T5268

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

MDL number:

biological source

goat

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

application(s)

indirect immunofluorescence: 1:32

conjugate

TRITC conjugate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Dong-Myung Kim et al.
Toxicological sciences : an official journal of the Society of Toxicology, 149(1), 4-14 (2015-09-20)
Drug toxicity is a key issue for drug R&D, a fundamental challenge of which is to screen for the targets genome-wide. The anticancer tyrosine kinase inhibitor sunitinib is known to induce cardiotoxicity. Here, to understand the molecular insights of cardiotoxicity...
Zeina Nasr et al.
BMC pharmacology & toxicology, 14, 58-58 (2013-11-14)
Chemotherapy-induced hair loss (alopecia) (CIA) is one of the most feared side effects of chemotherapy among cancer patients. There is currently no pharmacological approach to minimize CIA, although one strategy that has been proposed involves protecting normal cells from chemotherapy...
C Serradifalco et al.
European journal of histochemistry : EJH, 55(4), e41-e41 (2012-02-03)
During embryogenesis, the mammalian heart develops from a primitive heart tube originating from two bilateral primary heart fields located in the lateral plate mesoderm. Cells belongings to the pre-cardiac mesoderm will differentiate into early cardiac progenitors, which express early transcription...
Partial purification and immunohistochemical localization of ATP diphosphohydrolase from Schistosoma mansoni. Immunological cross-reactivities with potato apyrase and Toxoplasma gondii nucleoside triphosphate hydrolase.
Vasconcelos, et al.
The Journal of Biological Chemistry, 271, 22139-22145 (2021)
Sylvie François et al.
Journal of virology, 90(19), 8686-8697 (2016-07-22)
Varicella-zoster virus (VZV) is an alphaherpesvirus that causes varicella upon primary infection and zoster upon reactivation from latency in sensory ganglion neurons. The replication of herpesviruses requires manipulation of cell signaling pathways. Notably, CREB, a factor involved in the regulation...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service