T7188

Sigma-Aldrich

TFPI hydrochloride

NOS inhibitor

Synonym(s):
S-Ethyl-N-4-(trifluoromethyl)phenylisothiourea
Linear Formula:
C10H11N2SF3 · HCl
Molecular Weight:
284.73
MDL number:
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

Cl.CCSC(=N)Nc1ccc(cc1)C(F)(F)F

Gene Information

human ... NOS1(4842), NOS2(4843), NOS2B(201288), NOS2C(645740), NOS3(4846)

Biochem/physiol Actions

NOS inhibitor selective for neuronal isoform; Ki = 0.32 mM

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
B G Shearer et al.
Journal of medicinal chemistry, 40(12), 1901-1905 (1997-06-06)
S-Ethyl N-phenylisothiourea (4) has been found to be a potent inhibitor of both the human constitutive and inducible isoforms of nitric oxide synthase. A series of substituted N-phenylisothiourea analogues was synthesized to investigate the structure-activity relationship of this class of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.