All Photos(1)

T7878

Supelco

Tropine

Synonym(s):
3-Tropanol
Empirical Formula (Hill Notation):
C8H15NO
CAS Number:
Molecular Weight:
141.21
Beilstein:
80188
EC Number:
MDL number:

SMILES string

CN1[C@H]2CC[C@@H]1C[C@H](O)C2

InChI

1S/C8H15NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-8,10H,2-5H2,1H3/t6-,7+,8+

InChI key

CYHOMWAPJJPNMW-JIGDXULJSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service