MilliporeSigma
All Photos(1)

T9031

Supelco

Theophylline solution

drug standard, 1.0 mg/mL in methanol

MDL number:
PubChem Substance ID:

grade

drug standard

assay

≥98%

concentration

1.0 mg/mL in methanol

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

CN1C(=O)N(C)c2[nH]cnc2C1=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service