WH0000867M1

Sigma-Aldrich

Monoclonal Anti-CBL antibody produced in mouse

clone 6D12, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-CCBL, Anti-CBL2, Anti-Cas-Br-M (murine) ecotropic retroviral transforming sequence, Anti-RNF55
MDL number:
Pricing and availability is not currently available.

antibody form

purified immunoglobulin

clone

6D12, monoclonal

form

buffered aqueous solution

species reactivity

human

application(s)

indirect ELISA: suitable
western blot: suitable

isotype

IgG1κ

immunogen sequence

NIQSQAPSITESSTFGEGNLAAAHANTGPEESENEDDGYDVPKPPVPAVLARRTLSDISNASSSFGWLSLDGDPTTNVTEGSQVPERPPKPFPRRINSER (without GST)

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CBL(867)

Immunogen

CBL (NP_005179, a.a. 736-835) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.

Physical form

Solution in phosphate buffered saline, pH 7.2

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.