WH0005747M1

Sigma-Aldrich

Monoclonal Anti-PTK2 antibody produced in mouse

clone 2C3, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-FAK, Anti-FAK1, Anti-pp125FAK, Anti-PTK2 protein tyrosine kinase 2, Anti-FADK
MDL number:
NACRES:
NA.41
Pricing and availability is not currently available.

Quality Level

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C3, monoclonal

form

buffered aqueous solution

species reactivity

human

application(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

conjugate

unconjugated

GenBank® accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PTK2(5747)

General description

This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. At least four transcript variants encoding four different isoforms have been found for this gene, but the full-length natures of only two of them have been determined. (provided by RefSeq)

Immunogen

PTK2 (AAH28733, 355 a.a. ~ 490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EGFYPSPQHMVQTNHYQVSGYPGSHGITAMAGSIYPGQASLLDQTDSWNHRPQEIAMWQPNVEDSTVLDLRGIGQVLPTHLMEERLIRQQQEMEEDQRWLEKEERFLKPDVRLSRGSIDREDGSLQGPIGNQHIYQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Jinping Luo et al.
Biochemical and biophysical research communications, 450(3), 1212-1217 (2014-03-29)
Fertilization triggers rapid changes in intracellular free calcium that serve to activate multiple signaling events critical to the initiation of successful development. Among the pathways downstream of the fertilization-induced calcium transient is the calcium-calmodulin dependent protein tyrosine kinase PTK2b or...
Nadja Engel et al.
BMC complementary and alternative medicine, 14, 334-334 (2014-09-10)
Jatropha curcas (JCP1), Pyrenacantha staudtii (PS), Picralima nitida (ZI) and Jatropha gossypifolia (JCP2) are plants used in the African folklore for the treatment of various cancers. This study investigated the in vitro anticancer effects of the ethanol extracts against human...
Li Li et al.
Journal of molecular neuroscience : MN, 53(4), 580-589 (2014-01-09)
Glial cell line-derived neurotrophic factor (GDNF) is a potent neurotrophic factor for midbrain dopamine (DA) neurons, while the DA neurons in the ventral tegmental area (VTA) is a crucial part of the neural circuits associated with drug addiction. Recently, more...
D Barras et al.
Oncogene, 33(44), 5163-5172 (2013-11-12)
TAT-RasGAP(317-326), a peptide corresponding to the 317-326 sequence of p120 RasGAP coupled with a cell-permeable TAT-derived peptide, sensitizes the death response of various tumor cells to several anticancer treatments. We now report that this peptide is also able to increase...
M H A M van Miltenburg et al.
British journal of cancer, 110(11), 2747-2755 (2014-05-09)
Elevated expression of focal adhesion kinase (FAK) occurs in numerous human cancers including colon-, cervix- and breast cancer. Although several studies have implicated FAK in mammary tumour formation induced by ectopic oncogene expression, evidence supporting a role for FAK in...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.