WH0010598M1

Sigma-Aldrich

Monoclonal Anti-AHSA1 antibody produced in mouse

clone 1A2-A8, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-AHA1, Anti-p38, Anti-AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), Anti-C14orf3
MDL number:
NACRES:
NA.41

Quality Level

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A2-A8, monoclonal

form

buffered aqueous solution

species reactivity

human

application(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

conjugate

unconjugated

GenBank® accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... AHSA1(10598)

Immunogen

AHSA1 (AAH00321, 1 a.a. ~ 338 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAKWGEGDPRWIVEERANATNVNNWHWTERDASNWSTDKLKTLFLAVQVQNEEGKCEVTEVSKLDGEASINNRKGKLIFFYEWSVKLNWTGTSKSGVQYKGHVEIPNLSGENSVDEVEISVSLAKDEPDTNLVALMKEEGVKLLREAMGIYISTLKTEFTQGMILPTMNGESVDPVGQPALKTEERKAKPAPSKTQARPVGVKIPTCKITLKETFLTSPEELYRVFTTQELVQAFTHAPATLEADRGGKFHMVDGNVSGEFTDLVPEKHIVMKWRFKSWPEGHFATITLTFIDKNGETELCMEGRGIPAPEEERTRQGWQRYYFEGIKQTFGYGARLF

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

David M Taylor et al.
Cell stress & chaperones, 12(2), 151-162 (2007-08-11)
Induction of heat shock proteins (Hsps) is under investigation as treatment for neurodegenerative disorders, yet many types of neurons, including motor neurons that degenerate in amyotrophic lateral sclerosis (ALS), have a high threshold for activation of the major transcription factor...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service