MilliporeSigma
All Photos(6)

WH0114814M1

Sigma-Aldrich

Monoclonal Anti-GNRHR2 antibody produced in mouse

clone 4A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-GnRH receptor II 5TM, Anti-GnRHIIR, Anti-Type II GnRH receptor, Anti-gonadotropin-releasing hormone (type 2) receptor 2
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A5, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank® accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GNRHR2(114814)

General description

The receptor for gonadotropin releasing hormone 2 (GnRH2) is encoded by the GnRH2 receptor (GnRHR2) gene. In non-hominoid primates and non-mammalian vertebrates, GnRHR2 encodes a seven-transmembrane G-protein coupled receptor. However, in human, the N-terminus of the predicted protein contains a frameshift and premature stop codon. In human, GnRHR2 transcription occurs but whether the gene produces a functional C-terminal multi-transmembrane protein is currently unresolved. Alternative splice variants have been reported. An untranscribed pseudogene of GnRHR2 is also on chromosome 14. (provided by RefSeq)

Immunogen

GNRHR2 (NP_001457, 237 a.a. ~ 292 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TLGCRRGHQELSIDSSKEGSGRMLQEEIHAFRQLEVQKTVTSRRAGETKGISITSI

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service