Documents

XNAP2RE

Sigma-Aldrich

Extract-N-Amp Plant PCR Kit

sufficient for 1000 extractions, sufficient for 5000 amplifications

Pricing and availability is not currently available.

usage

sufficient for 1000 extractions
sufficient for 5000 amplifications

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. A novel Extraction Solution eliminates the need for conventional freezing of plant tissues with liquid nitrogen, mechanical disruption, organic extraction, column purification, or precipitation of DNA.

Features and Benefits

  • Single-step extraction of plant genomic DNA for PCR in less than 15 minutes
  • No freezing, mechanical disruption, organic extraction, column purification or precipitation required
  • Specially formulated PCR ReadyMix for use with extract
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • REDExtract-N-Amp requires no loading buffers or tracking dyes required for gel analysis
  • Compatible with high-throughput requirements for genetic analysis of plants
  • Extract stable at 4 °C for at least 6 months

Components

Kits contain Dilution solution, Extraction solution, and Extract-N-Amp PCR Reaction Mix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.