Documents
Z227471
Millipore
Millex® syringe filter units, disposable, PTFE
pore size 0.45 μm, diam. 4 mm, fluoropore PTFE membrane
NACRES:
NB.24
Pricing and availability is not currently available.
material
PTFE membrane
fluoropore PTFE membrane
high-density polyethylene housing
sterility
non-sterile
packaging
pack of 100 ea
mfr. no.
Millipore, SLFHR04NL
diam.
4 mm
volume
≤1 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
General description
All units contain PTFE filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings.
Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 mL; the 13 mm units are recommended for volumes of 0.5-10 mL.
Application
HPLC; organic solvents and solutions
Legal Information
Millex is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.