Z355879

Horizontal IBI electrophoresis units

Model QS 710, Quick Screening

NACRES:
NB.22

feature

capacity 30 samples two 15-tooth combs
comb positions 2

manufacturer/tradename

IBI IB51000

buffer capacity

250 mL

gel W × L

7 cm × 10 cm

Looking for similar products? Visit Product Comparison Guide

General description

includes two 1.5mm × 8-tooth combs
These units are complete designs that incorporate the best features of the popular IBI units.

Features:
  • Durable acrylic construction with fewer seams and more unitized parts
  • Vented safety lid allows heat to escape and improves visibility by reducing condensation
  • External casting system allows one gel to be cast (without tape) while another is running
  • Electrical contact is made by gently placing the lid on the chamber, eliminating sample disturbance during set-up
  • Gel trays are transparent to 312 nm UV light to allow visualization on the tray
  • Dark stripes below each comb position make it easy to see wells in submerged gel
  • Platinum electrodes and gold-plated contacts resist corrosion

Packaging

Each of these units comes complete and includes a casting system with UV-transparent gel tray, leveling bubble, power leads, 2 combs and buffer recirculating ports. Does not include extra gel-casting accessories, combs, or replacement parts.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service