Documents
Z372536
Sigma-Aldrich
Combs for vertical mini-gel, 11.3 cm wide
size 0.75 mm × 10 wells
Pricing and availability is not currently available.
size
0.75 mm × 10 wells
General description
The following components are necessary to construct one complete single gel casting assembly: one set of glass plates, one set of spacers, one gasket, one comb, and gel casting clamps (one set of three for mini-size gel; two sets of three for standard size gels). Be sure to select the same length glass plates, spacers and gaskets; the same thickness spacers, comb and gasket.
For dual gel casting, order two of each component, but substitute dual gel casting clamps for the single gel casting clamps. For the second dimension of 2D electrophoresis, spacers, gaskets and a "1-well" comb must match the thickness of the first dimension tube gel.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.