All Photos(1)
Z374512
Sigma-Aldrich
Brinkmann bottle-top dispenser
1.0-5.0 mL
availability
not available in EU
measuring range
1.0-5.0 mL
General description
Brinkman bottletop dispensers offer a unique combination of features:
  • PTFE "wiping" piston displaces reagent from the cylinder without leaving any residue behind, eliminating piston swelling and freezing
  • Grooved positive lock volume adjustment
  • Unique telescoping filling tube
  • Individual calibration certificate
  • Completely autoclavable without disassembly
  • Clear plastic sleeve surrounds and protects the glass cylinder
Accuracy ±0.5% (±0.6% for 0.1-2.5 mL size). Reproducibility of 0.1%.
Adapters and accessories allow connection to almost any size reagent bottle. Dispensers with maximum volume of 2.5 to 10 mL connect directly to bottles with 33 mm threaded neck and are supplied with 28 mm, 38 mm, and 45 mm adapters. Dispensers with maximum volumes of 25 to 100 mL connect directly to bottles with 45 mm threaded neck and are supplied with 33 mm and 38 mm adapters
Certificate of Analysis
Certificate of Origin