All Photos(1)
Z379441
Sigma-Aldrich
Hanna general purpose combination pH electrode
Glass body (Double junction refillable)
description
BNC connector
Double junction Ag/AgCI
manufacturer/tradename
Hanna HI 1043B
pH measuring range
0-14
temp measuring range
−5-100 °C
L
120 mm , Overall
General description
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).