All Photos(1)
Z380369
Nalgene® Griffin beakers
capacity 600 mL, translucent polypropylene
material
translucent polypropylene
packaging
pkg of 4
manufacturer/tradename
Nalgene® 1201
availability
not available in EU
capacity
600 mL
General description
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).