Documents
Z649120
Radiation B-Safe bin box bags
W × D × H 280 mm × 280 mm × 510 mm
material
polyethylene
manufacturer/tradename
Jencons
availability
not available in USA and Canada
W × D × H
280 mm × 280 mm × 510 mm
General description
Supplied in benchtop dispenser boxes. Heat sealed for leak protection. Each bag is box shaped and has a double string neck pull. Printed with black radiation warning symbol on a yellow background with maximum fill line.
Certificate of Analysis
Certificate of Origin