Documents
Z717029
Corning® CellBIND® Surface CellSTACK® cell culture chambers
Pricing and availability is not currently available.
description
1 stack
mfr. no.
Corning 3330
surface area
636 cm2
suitability
suitable for (scale-up cell culture)
General description
The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.
Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.
CellSTACK chambers are ideal for cell culture scale-up, providing cell yields in the 107 - 109 range and are useful alternatives to multiple roller bottles or spinner flasks.
  • Ideal for cell culture scale-up
  • useful alternatives to multiple roller bottles or spinner flasks
  • High mechanical strength and structural integrity
  • 100% leak tested
  • Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
  • Easy to use larger openings with threaded closures and vented caps
  • Filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures
Legal Information
CellBIND is a registered trademark of Corning, Inc.
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.