13071-U

Supelco

PTFE Packed GC Column (General Configuration)

phase none, matrix 80/100 Porapak QS support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8  in. × 2.1 mm

material

PTFE

parameter

0-250 °C temperature (isothermal or programmed)

application(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

6.0 ft (1.8 m) × 1/8  in. × 2.1 mm

matrix

80/100 Porapak QS support

matrix active group

none phase

column type

packed GC

compatibility

configured for General Use

Looking for similar products? Visit Product Comparison Guide

General description

In a packed column, the carrier gas percolates through the porous bed and separation is achieved by moving the solutes apart. PTFE packing is fibrous and is made from machine-braided yarn. PTFE-based packing material is resistant over the range 0-14 pH and fluid temperature upto 258 °C. It also reduces friction during pump application. PTFE is limited to rotational surface speeds below 0.5 m/s due to its poor thermal conductivity and high thermal expansion coefficient. Porapak QS is a cross-linked polyaromatic resin which is capable to extract organic compounds and any trace metals associated with organic molecules.
For low temperature applications, a PTFE column offers the flexibility of metal with inertness approaching that of glass. This column is of a general configuration, and can be carefully bent to fit most instruments.

Certificate of Analysis

Certificate of Origin

Principles and Applications
Heinz K. Muller & Bernard S. Nau
Fluid Sealing Technology, 200-201 (1998)
Adsorption Behavior of Transition Metal Complexes on Polychlorotrifluoroethylene Resin
Chiyoshi AKITA
Analytical Sciences, 3, 473-474 (1987)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service