All Photos(1)
13992-U
Supelco
Packed GC Column Set
suitable for 2261 per GPA (5-column set, pretested)
Agency
suitable for GPA 2261 (5-column set, pretested)
description
Pretested with a C1-C6+ hydrocarbon mix
technique(s)
gas chromatography (GC): suitable
column type
packed GC
General description
This column set is specifically designed for a specific column switching application. Column manufacturing procedurs have been optimized resulting in column sets that provide highly reproducible analyses.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).