Documents

20361

Supelco

Carbotrap® 201

for use with Dynatherm 850/890 TDU, I.D. × O.D. × L 1 mm × 6 mm × 4.5 in., glass TD tube (focusing tube)

Synonym(s):
Carbotrap 201 Dynatherm 890, 850

material

glass TD tube (focusing tube)

packaging

pkg of 1 ea

I.D. × O.D. × L

1 mm × 6 mm × 4.5 in.

matrix

Carbotrap® 201

Featured Industry

Environmental
Industrial Hygiene

compatibility

for use with Dynatherm 850/890 TDU

Looking for similar products? Visit Product Comparison Guide

Application

focusing semivolatile to very volatile compounds

Legal Information

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
Dynatherm is a trademark of CDS Analytical

WGK Germany

-

Certificate of Analysis
Certificate of Origin
Trine Salomón et al.
Cell chemical biology, 24(8), 935-943 (2017-08-19)
The α-oxoaldehyde methylglyoxal is a ubiquitous and highly reactive metabolite known to be involved in aging- and diabetes-related diseases. If not detoxified by the endogenous glyoxalase system, it exerts its detrimental effects primarily by reacting with biopolymers such as DNA...
S Ghittori et al.
Giornale italiano di medicina del lavoro, 10(4-5), 201-205 (1988-07-01)
A laboratory study, using generated atmosphere containing 0.14/23.3 mg/m3 of Benzene, was conducted to adapt an existing industrial hygiene monitoring method for measuring low concentrations of Benzene. This method was developed to determine concentrations of Benzene in the ambient air...
Florian Gahleitner et al.
Bioanalysis, 5(18), 2239-2247 (2013-09-24)
In-community non-invasive identification of asthma-specific volatile organic compounds (VOCs) in exhaled breath presents opportunities to characterize phenotypes, and monitor disease state and therapies. The feasibility of breath sampling with children and the preliminary identification of childhood asthma markers were studied....
Yifei Sun et al.
Journal of environmental sciences (China), 25(1), 213-219 (2013-04-17)
A group parameter approach using "total organic halogen" is effective for monitoring gaseous organic halogen compounds, including fluorine, chlorine, and bromine compounds, generated from combustion. We described the use of barrier-discharge radiofrequency-helium-plasma/atomic emission spectrometry, for the detection of semi- and...
M L Fiorentino et al.
La Medicina del lavoro, 81(2), 107-118 (1990-03-01)
Benzene is a widely diffuse solvent (atmosphere, cigarette smoke, some foods); in the industrial environment benzene is currently present at concentrations of ppm. A valid method of biological monitoring that is easy to perform is needed for assessing occupational and...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.