All Photos(1)

20912-U

Supelco

Carbotrap® 300

glass TD tube, O.D. × I.D. × L 1/4  in. × 4 mm × 7 in., preconditioned, pkg of 1 ea

Synonym(s):
Carbotrap® 300 glass TD tube 7IN X 1/4IN for Tekmar®

material

glass TD tube

Agency/Method

EPA TO-17

description

Sealed with TDS³ Storage Container

feature

preconditioned

packaging

pkg of 1 ea

O.D. × I.D. × L

1/4  in. × 4 mm × 7 in.

matrix

Carbosieve® S-III carbon molecular sieve (CMS)
Carbotrap® 300 (Packed with Carbotrap C, Carbotrap B, Carbosieve SIII)
Carbotrap® C graphitized carbon black (GCB)
Carbotrap® graphitized carbon black (GCB)

Featured Industry

Environmental
Industrial Hygiene

compatibility

for use with Tekmar® AEROTrap 6000 models

Looking for similar products? Visit Product Comparison Guide

Legal Information

Carbosieve is a registered trademark of Sigma-Aldrich Co. LLC
Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
Tekmar is a registered trademark of Tekmar Co.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service