All Photos(1)

21059-U

Supelco

Purge/Trap A

for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT3

Synonym(s):
PURGE TRAP A, Tekmar®
NACRES:
NB.21

Quality Level

Featured Industry

Environmental

compatibility

for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT3

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Tekmar is a registered trademark of Tekmar Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service