All Photos(1)
22744
Supelco
Fritted Purge Sampler
size 25 mL, right stem, Tekmar® 14-3546-124
material
glass
Quality Level
feature
right stem
size
25 mL
Featured Industry
Industrial Hygiene
compatibility
for use with Tekmar® 2000/3000
Features and Benefits
Note: All 1/4 in. O.D. glass ends are ground glass.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Certificate of Analysis
Certificate of Origin