MilliporeSigma
All Photos(1)

28956-U

Supelco

Carbosieve SIII

glass TD tube, unconditioned

material

glass TD tube

Agency

EPA TO-2

feature

unconditioned

packaging

pkg of 10 ea

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Carbosieve S-III carbon molecular sieve

application(s)

environmental

compatibility

for use with PerkinElmer ATD-50,ATD-400 and TurboMatrix (TMX) Series

Looking for similar products? Visit Product Comparison Guide

Legal Information

Carbosieve is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service