All Photos(1)

442680

Supelco

Hexatriacontane

analytical standard

Linear Formula:
CH3(CH2)34CH3
CAS Number:
Molecular Weight:
506.97
Beilstein:
1712367
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

vapor density

17.6 (vs air)

form

neat

packaging

ampule of 1000 mg

application(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

265 °C/1 mmHg (lit.)

mp

74-76 °C (lit.)

Featured Industry

Environmental
Petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C36H74/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-36H2,1-2H3

InChI key

YDLYQMBWCWFRAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service