442706

Supelco

Tetracontane

analytical standard, Product 442706 has been discontinued. Product 87086-250mg can be used as a replacement.

Synonym(s):
n-Tetracontane
Linear Formula:
CH3(CH2)38CH3
CAS Number:
Molecular Weight:
563.08
Beilstein/REAXYS Number:
1800152
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

form

neat

packaging

ampule of 500 mg

application(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

80-84 °C (lit.)

Featured Industry

Environmental
Petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C40H82/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3

InChI key

KUPLEGDPSCCPJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetracontane belongs to the class of saturated hydrocarbons.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tetracontane may be used as an analytical reference standard for the determination of tetracontane in:
  • Ambient aerosol samples and in aerosol filter samples by an in-injection port thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS).
  • Soil and water samples by GC equipped with flame ionization detector (FID).

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Analysis of insect cuticular hydrocarbons using matrix-assisted laser desorption/ionization mass spectrometry.
Cvacka J, et al.
Journal of Chemical Ecology, 32(2), 409-434 (2006)
Challenges in the determination of petroleum hydrocarbons in water by gas chromatography (hydrocarbon index).
Drozdova S, et al.
Fuel: The Science and Technology of Fuel and Energy, 113(2), 527-536 (2013)
Precautions for in-injection port thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS) as applied to aerosol filter samples.
Ho SSH, et al.
Atmospheric Environment, 45(7), 1491-1496 (2011)
Evaluation of an in-injection port thermal desorption-gas chromatography/mass spectrometry method for analysis of non-polar organic compounds in ambient aerosol samples.
Ho SSH, et al.
Journal of Chromatography A, 1200(2), 217-227 (2008)
Effect of sample matrix on the determination of total petroleum hydrocarbons (TPH) in soil by gas chromatography?flame ionization detection.
Saari E, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 87(2), 113-118 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.