48134

Supelco

MISA Group 19 Base-Neutral Extractables Mix A

2000 μg/mL each component in dichloromethane

grade

analytical standard

Quality Level

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bis(2-chloroethoxy)methane
    Bis(2-chloroethyl) ether
    4-Bromodiphenyl ether
    Camphene
    4-Chlorodiphenyl ether
    Bis-(2-chloroisopropyl) ether
    1-Chloronaphthalene
    2-Chloronaphthalene
    Diphenylamine
    1-Methylnaphthalene
    2-Methylnaphthalene
    5-Nitroacenaphthene
    See All (12)

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service