MilliporeSigma
All Photos(1)

48824

Supelco

Trihalomethanes Kit

analytical standard

grade

analytical standard

Agency

EPA 601

feature

standard type calibration

packaging

pkg of 1 kit

application(s)

environmental

storage temp.

2-8°C

Kit Components Only

Product No.
Description

  • Trihalomethanes Calibration Mix (48746) 1 mL

  • Bromoform (48618) 1 mL

  • Bromodichloromethane (48615) 1 mL

  • Dibromochloromethane (48608) 1 mL

  • Chloroform (48603) 1 mL

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service