All Photos(1)
508608
Supelco
Septa, PTFE/Silicone
blue PTFE/silicone, diam. × thickness 20 mm × 0.105 in., hardness 45 (Durometer: shore A), pkg of 100 ea
material
PTFE face
blue PTFE/silicone
silicone body
hardness
45 (Durometer: shore A)
packaging
pkg of 100 ea
manufacturer/tradename
Chromacol 20-ST3
parameter
250 °C temp. limit
O.D.
20 mm
diam. × thickness
20 mm × 0.105 in.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).