MilliporeSigma
All Photos(1)

51243-U

Upchurch Mini MicroFilter Assembly

pore size 1 μm, titanium frit, pkg of 1 ea

NACRES:
SB.52

packaging

pkg of 1 ea

pore size

1 μm

Looking for similar products? Visit Product Comparison Guide

General description

Ultra-low swept volume - 0.85 μL. Use with Microtight tubing sleeves for capillary tubing. Available with 1 μm or 2 μm frit capsules (stainless steel frit in color-coded PEEK capsule). 57421-U includes 5, 2μm filter capsules, 2 microferrules, and 2 female natural PEEK nuts and catalog number 54722-U includes 5, 1μm filter capsules, 2 microferrules, and 2 female natural PEEK nuts. 51242-U includes 5, 1 μm nano filter capsule (PEEK with stainless steel frit), 2, microferrule for 360 μm O. D. tubing, 2 female nuts. 51243-U ncludes 5, 1 μm nano filter capsule (PEEK with titanium frit), 2, microferrule for 360 μm O. D. tubing, 2 female nuts.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service