Documents
56080-U
Omnifit® Tube Endfittings
for use with 1/8 in. O.D. tubing, mixed polypropylene
material
mixed polypropylene
packaging
pkg of 10 ea
fitting
1/4-28 male UNF nut (Gripper Fitting required)
compatibility
for use with 1/8 in. O.D. tubing
General description
Each endfitting has an internal taper to hold the tubing centrally; includes a lock nut for panel mounting.
Gripper fitting required (purchase separately):
for 1/16in. O.D. tubing p/n 57417
for 1/8in. O.D. tubing p/n 57418
Legal Information
Omnifit is a registered trademark of Diba Industries
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.