All Photos(1)

59454

Opti-Max® Inlet Check Valve

stainless steel, for use with 100A, 110A/B, 112, 112M, 114, 114M, 116, 118, 125, 126, 127, 128, Beckman 240720

Synonym(s):
Optimize Technologies HPLC Pump Part for Beckman®

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Beckman 240720

compatibility

for use with 100A, 110A/B, 112, 112M, 114, 114M, 116, 118, 125, 126, 127, 128

Looking for similar products? Visit Product Comparison Guide

Legal Information

Beckman is a registered trademark of Beckman Coulter, Inc.
Opti-Max is a registered trademark of Optimize Technologies, Inc.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service