868091

Supelco

QFT-2 Standard

in heptane, analytical standard

EC Number:

grade

analytical standard

packaging

pkg of 100 mL

concentration

in heptane

application(s)

HPLC: suitable
gas chromatography (GC): suitable

format

single component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Phenanthrene 4 μg/mL
    p-Xylene 200 μg/mL

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service