16851

Sigma-Aldrich

Atto 665 (free acid)

≥95.0% (HPCE)

NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

assay

≥95.0% (HPCE)

mol wt

Mw 723 g/mol

fluorescence

λex 663 nm; λem 684 nm±5 nm in 0.1 M phosphate pH 7.0

storage temp.

−20°C

General description

Atto 665 is a new fluorescent label closely related to Atto 647N. It shows extraordinary high fluorescence quantum yield, excellent thermal and photo-stability, outstanding ozone resistance, and very little triplet formation. Atto 665 is a cationic (charge +1). The dye is moderately hydrophilic.

find more information
here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Articles
Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.
Read More
Related Content
Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.