44756

Sigma-Aldrich

Atto 580 Q NHS ester

≥90% (coupling to amines)

MDL number:
Pricing and availability is not currently available.

Quality Level

assay

≥90% (coupling to amines)

application(s)

UV/Vis spectroscopy: suitable

fluorescence

λex 586 nm

suitability

corresponds for UV test

storage temp.

−20°C

Application

Atto 580Q is a new quencher for fluorescence characterized by high molecular absorption (110.000). Atto 580Q is characterized by a high photostability and thermostability. It is moderately hydrophilic, and soluble in polar solvents as DMF or DMSO. Atto 580 Q NHS ester is used to label amines in proteins and amino modified oligonucleotides.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.