68538

Sigma-Aldrich

Atto Rho13 NHS ester

NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

fluorescence

λex 600 nm; λem 625 nm

storage temp.

−20°C

Application

Atto Rho13 NHS ester is a new fluorescent label belonging to the class of rhodamines. Characteristic features of the label are strong absorption, high fluorescence quantum yield, and high thermal and photo-stability. ATTO Rho13 is a cationic dye. After coupling to a substrate the dye carries a net electrical charge of +1.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

S R Kwon et al.
International journal of cosmetic science, 34(1), 97-101 (2011-09-29)
Enamel and dentin are susceptible to extrinsic and intrinsic stains. The purposes of this study were to determine the penetration pattern of Rhodamine B and dextran-conjugated Rhodamine B into the enamel and dentin as observed by confocal laser microscopy and...
Elizabeth S Childress et al.
Analytical chemistry, 84(3), 1235-1239 (2012-01-28)
We present dye-doped polymer nanoparticles that are able to detect mercury in aqueous solution at parts per billion levels via fluorescence resonance energy transfer (FRET). The nanoparticles are prepared by reprecipitation of highly fluorescent conjugated polymers in water and are...
Melanie Kucki et al.
Journal of the Royal Society, Interface, 9(69), 727-733 (2011-08-26)
The incorporation of rhodamine dyes in the cell wall of diatoms Coscinodiscus granii and Coscinodiscus wailesii for the production of luminescent hybrid nanostructures is investigated. By systematic variation of the substitution pattern of the rhodamine core, we found that carbonic...
Yasmine A Gomaa et al.
International journal of pharmaceutics, 438(1-2), 140-149 (2012-09-11)
Drug flux across microneedle (MN)-treated skin is influenced by the characteristics of the MN array, formed microconduits and physicochemical properties of the drug molecules in addition to the overall diffusional resistance of microconduits and viable tissue. Relative implication of these...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.