68761

Sigma-Aldrich

Atto Rho12 NHS ester

reagent grade, suitable for fluorescence

NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

grade

reagent grade

fluorescence

λex 576 nm; λem 601 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho12 NHS ester is a new rhodamine dye for application in life sciences, e.g. labeling of DNA, RNA or proteins. The label shows strong absorption, high fluorescence quantum yield, and high photostability.
After coupling to a substrate Atto Rho12 NHS ester carries a net electrical charge of +1.
The dye is moderately hydrophilic. Atto Rho12 NHS ester consists of a mixture of three isomers with practically identical absorption and fluorescence properties.

find more information
here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.