72763

Sigma-Aldrich

Atto Rho12 maleimide

Pricing and availability is not currently available.

fluorescence

λex 576 nm; λem 601 nm

storage temp.

−20°C

General description

Atto Rho12 is a new rhodamine dye for application in life sciences, e.g. labelling of DNA, RNA or proteins. The label shows strong absorption, high fluorescence quantum yield, and high photostability. After coupling to a substrate
Atto Rho12 carries a net electrical charge of +1.
The dye is moderately hydrophilic. Atto Rho12 consists of a mixture of three isomers with practically identical absorption and fluorescence properties.

find more information
here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Codes

Xi

Risk Statement

36/38

Safety Statement

26

RIDADR

NONH for all modes of transport

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.